Order Entry
Norway
ContactUsLinkComponent
 
2-Propyl-4-(4-pyridylazo)phenol ≥98.0% (by HPLC)
  TCIAP1442-5G
 :  TCI
 :  
2-Propyl-4-(4-pyridylazo)phenol ≥98.0% (by HPLC)
  TCIAP1442-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Yellow to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    175.0 to 179.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :241.29 g/mol
 :253124-08-2
 :C₁₄H₁₅N₃O
 :MFCD06797155