1-Pyrenemethanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:24463-15-8
:
Formula:C₁₇H₁₂O
MDL Number:MFCD00029252
:232.28 g/mol
Specification Test Results
- Appearance:White to Amber powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:123.0 to 127.0 deg-C
- Solubility in hot Toluene:almost transparency