Pullulan
Supplier: TCI
Certificates
About this item
CAS Number:9057-02-7
:
Formula:(C₆H₁₂O₅)n
MDL Number:MFCD00081940
EINECS:232-945-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Total Nitrogen:max. 0.05 %
- Drying loss:max. 8.0 %
- Ignition residue (Sulfate):max. 5.0 %
- Viscosity:15.0 to 180.0 mPa-s (10%, H2O, 30 deg-C)