O-Phospho-L-tyrosine ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAP0959-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Specific rotation [a]20/D:-5.0 to -8.0 deg (C=1, 2mol/L HCl)
Specification Test Results
About this item
:
:261.17 g/mol
Storage Temperature:Freezer
CAS Number:21820-51-9
Formula:C₉H₁₂NO₆
MDL Number:MFCD00002603