
Ponceau Xylidine
Catalog # TCIAP0590-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Red to Dark red to Brown powder to crystal
- Lambda max.:504.0 to 508.0 nm (H2O)
- Absorbance (E1%1cm):min. 490 (H2O, 504.0 to 508.0 nm) (calcd.on dried substance)
- Drying loss:max. 12.0 %
Specification Test Results
About this item
:
:480.42 g/mol
EINECS:223-178-3
:GHS08
CAS Number:3761-53-3
Formula:C₁₈H₁₆N₂O₇S₂
MDL Number:MFCD00003897