Phenyl-1-Propenyl Ketone ≥80.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:495-41-0
:
Formula:C₁₀H₁₀O
MDL Number:MFCD00059386
:146.19 g/mol
Boiling Pt:122°C
EINECS:207-800-0
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 80.0 %