Order Entry
Norway
ContactUsLinkComponent
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether ≥98.0% (by GC)
Flammable
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
1H,1H,5H-Octafluoropentyl 1,1,2,2-tetrafluoroethyl ether ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results