Monoethyl-4-oxoheptanedioate ≥97.0% (by titrimetric analysis)
Catalog # TCIAO0211-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:69.0 to 72.0 deg-C
Specification Test Results
About this item
:
:202.21 g/mol
:GHS07
CAS Number:1506-55-4
Formula:C₉H₁₄O₅
MDL Number:MFCD00059093