3-Nitro-L-tyrosine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:621-44-3
:
Formula:C₉H₁₀N₂O₅
MDL Number:MFCD00007123
:226.3 g/mol
Melting Pt:224°C
EINECS:210-688-6
Specification Test Results
- Appearance:White to Amber powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+2.0 to +4.0 deg (C=1、1mol/LHCl)