L-Norvaline ethyl ester hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:40918-51-2
:
Formula:C₇H₁₅NO₂
MDL Number:MFCD08458628
:181.66 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:108.0 to 117.0 deg-C
- Specific rotation [a]20/D:+8.0 to +10.0 deg (C=2, H2O)