5-Norbornen-2-yl acetate (endo- and exo- mixture) ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:6143-29-9
:
Formula:C₉H₁₂O₂
MDL Number:MFCD00167565
:152.19 g/mol
Boiling Pt:73°C
Flash Pt:62°C
EINECS:228-144-1
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 96.0 % (total of isomers)