

8-Nitro-7-quinolinecarboxaldehyde ≥98.0% (by GC, titration analysis)
Catalog # TCIAN0852-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:178.0 to 182.0 deg-C
Specification Test Results
About this item
:
:202.17 g/mol
:GHS07
:GHS08
CAS Number:101327-87-1
Formula:C₁₀H₆N₂O₃
MDL Number:MFCD08436438