Order Entry
Norway
ContactUsLinkComponent
 
Sodium-4-nitrophenylacetate ≥99.0% (by titrimetric analysis)
  TCIAN0391-25G
 :  TCI
 :  
Sodium-4-nitrophenylacetate ≥99.0% (by titrimetric analysis)
  TCIAN0391-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Nonaqueous Titration):
    min. 99.0 %

 

 

 :
 :203.13 g/mol
 :7063-24-3
 :C₈H₇NO₄
 :MFCD00070537