1-Hexyloxy-4-nitrobenzene ≥98.0% (by GC)
Catalog # TCIAN0373-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White or Colorless to Brown powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Freezing point:23.0 to 26.0 deg-C
Specification Test Results
About this item
:
:223.27 g/mol
CAS Number:15440-98-9
Formula:C₁₂H₁₇NO₃
MDL Number:MFCD00043610