5-Nitro-2-furaldehyde diacetate ≥97.0% (by GC)
Catalog # TCIAN0218-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:87.0 to 92.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:243.17 g/mol
EINECS:202-166-1
CAS Number:92-55-7
Formula:C₉H₉NO₇
MDL Number:MFCD00003244