4-Mercapto-4-methyl-2-pentanone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19872-52-7
:
Formula:C₆H₁₂OS
MDL Number:MFCD00085208
Boiling Pt:57°C
Flash Pt:54°C
EINECS:243-386-8
UN:3336
ADR:3,III
:132.23 g/mol
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Iodometric Titration):min. 98.0 %