2'-C-Methyluridine ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:31448-54-1
:
Formula:C₁₀H₁₄N₂O₆
MDL Number:MFCD02682960
:258.23 g/mol
Melting Pt:112°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Specific rotation [a]20/D:+80.0 to +84.0 deg (C=1, H2O)
Specification Test Results
Frequently Bought Together