

Mitoxantrone ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:65271-80-9
:
Formula:C₂₂H₂₈N₄O₆
MDL Number:MFCD00242942
:444.49 g/mol
Merck Index:14,06217
UN:3077
ADR:9,III
:GHS08
:GHS09
Storage Temperature:Freezer
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:170.0 to 174.0 deg-C