m-PEG12-Mal ≥95.0% (by HPLC)
TCIAM3051-25MG
: TCI
:
- Pk:25 mg
- Seq:2315
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):3.50 to 4.50 %
:
:710.807 g/mol
:Freezer
:88504-24-9
:C₃₂H₅₈N₂O₁₅
:MFCD11041152