Methyl-PEG12-Azide ≥98.0% (by HPLC)
TCIAM3049-25MG
: TCI
:
- Fp:25 mg
- Seq:2315
- Appearance:Colorless to Light yellow clear liquid
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):6.50 to 7.80 %
:
:585.69 g/mol
:Fryseskap
:2170098-29-8
:C₂₅H₅₁N₃O₁₂