Boc-Asp-OMe ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAM2970-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:87.0 to 91.0 deg-C
- Specific rotation [a]20/D:-20.0 to -23.0 deg (C=1, MeOH)
Specification Test Results
About this item
:
:247.247 g/mol
CAS Number:98045-03-5
Formula:C₁₀H₁₇NO₆
MDL Number:MFCD03094777