
1-Methyl-3-phenyl-2-thiourea ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:2724-69-8
:
Formula:C₈H₁₀N₂S
MDL Number:MFCD00022130
:166.25 g/mol
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:110.0 to 114.0 deg-C