Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-bromo-2-fluorobenzoate
Methyl-3-bromo-2-fluorobenzoate ≥98.0% (by GC)
Methyl-3-bromo-2-fluorobenzoate ≥98.0% (by GC)
:
TCI
Methyl-3-bromo-2-fluorobenzoate ≥98.0% (by GC)
:
TCI
:
206551-41-9
:
:
C₈H₆BrFO₂
:
MFCD09261255
:
233.04 g/mol
:
GHS07
Appearance:
White to Almost white powder to crystal
Purity (GC):
min. 98.0 %
Melting point:
35.0 to 39.0 deg-C