2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:168425-64-7
:
Formula:C₁₆H₁₅N₃O₂
MDL Number:MFCD10568147
:281.32 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:211.0 to 215.0 deg-C