
6'-Methoxy-2,3'-bipyridine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:381725-49-1
:
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD11707142
:186.21 g/mol
Boiling Pt:104°C
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 97.0 %