Fumaric acid monomethyl ester ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2756-87-8
:
Formula:C₅H₆O₄
MDL Number:MFCD00063174
:130.1 g/mol
EINECS:220-412-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:145.0 to 149.0 deg-C