Methylamine ∼7% ∼2 mol/L in THF
Supplier: TCI
Certificates
About this item
CAS Number:74-89-5
:
Formula:CH₅N
MDL Number:MFCD00008104
:31.06 g/mol
Merck Index:14,06014
Storage Temperature:Refrigerator
EINECS:200-820-0
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):7.0 to 9.0 w/w%