Methyl-ɑ-bromophenylacetate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3042-81-7
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00013535
:229.07 g/mol
Boiling Pt:172°C
Flash Pt:150°C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %