

10-Methyl-9-phenylacridinium perchlorate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:36519-61-6
:
Formula:C₂₀H₁₆ClNO₄
MDL Number:MFCD09038537
:369.80 g/mol
UN:1479
ADR:5.1,III
:GHS03
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %