kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
2-Amino-1-methoxybutane
2-Amino-1-methoxybutane ≥90.0% (by GC)
2-Amino-1-methoxybutane ≥90.0% (by GC)
:
TCI
2-Amino-1-methoxybutane ≥90.0% (by GC)
:
TCI
:
63448-63-5
:
:
C₅H₁₃NO
:
MFCD00047920
:
103.17 g/mol
:
264-149-5
:
2734
:
8,II
:
GHS02
:
GHS05
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 90.0 %