Order Entry
Norway
ContactUsLinkComponent
1-(2,5-Dichloro-4-sulphophenyl)-3-methyl-5-pyrazolone monohydrate ≥98.0% (by titrimetric analysis)
1-(2,5-Dichloro-4-sulphophenyl)-3-methyl-5-pyrazolone monohydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0663-25G
Supplier:  TCI
CAS Number:  
1-(2,5-Dichloro-4-sulphophenyl)-3-methyl-5-pyrazolone monohydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0663-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    5.0 to 6.0 %

Specification Test Results

About this item

 :
 :323.14 g/mol
EINECS:201-541-7
 :GHS07
CAS Number:84-57-1
Formula:C₁₀H₈Cl₂N₂O₄S
MDL Number:MFCD00035706