Mucicarmine
Catalog # TCIAM0468-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Lambda max.:540.0 to 550.0 nm (H2O)
- Absorbance (E1%1cm):min. 50 (H2O, 540 to 550 nm)
Specification Test Results
About this item
:
EINECS:257-173-2
CAS Number:51395-97-2
MDL Number:MFCD00071289