
2,2'-Methylenebis(6-tert-butyl-4-ethylphenol) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:88-24-4
:
Formula:C₂₅H₃₆O₂
MDL Number:MFCD00026296
:368.56 g/mol
EINECS:201-814-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:124.0 to 128.0 deg-C
- Solubility in Methanol:almost transparency