
3-Methyl-2-benzothiazolinonehydrazone hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4338-98-1
:
Formula:C₈H₁₀ClN₃S
MDL Number:MFCD00005816
:215.70 g/mol
EINECS:224-393-5
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 % (after drying)
- Solubility in hot Water:almost transparency
- Drying loss:max. 2.0 %