
Monosodium maleate trihydrate ≥98.0% (by titrimetric analysis)
TCIAM0016-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:26.0 to 30.0 %
:
:138.05 g/mol
:221-461-6
:GHS07
:3105-55-3
:C₄H₄O₄
:MFCD00070246