Levodropropizine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:99291-25-5
:
Formula:C₁₃H₂₀N₂O₂
MDL Number:MFCD00866852
:236.32 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:103.0 to 107.0 deg-C
- Specific rotation [a]20/D:-24.0 to -28.0 deg (C=1, CHCL3)