Lactobionic acid ≥96.0% (by titrimetric analysis) (mixture of Acid form and Lactone form)
: TCI
:96-82-2
:
:C₁₂H₂₂O₁₂
:MFCD00078147
:358.30 g/mol
:14,05342
:202-538-3
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):96.0 to 106.0 %
- Specific rotation [a]20/D:+21.0 to +27.0 deg (C=8.8, H2O) (after 4hrs)
Frequently Bought Together