
Isopentylamine hydrobromide ≥98.0% (by titrimetric analysis)
: TCI
:2733412-57-0
:
:C₅H₁₃N·HBr
:MFCD33021450
:168.08 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together