
Isobutylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:74098-36-5
:
Formula:C₄H₁₁N·HBr
MDL Number:MFCD29089367
:154.05 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together