Order Entry
Norway
ContactUsLinkComponent
 
(R,R)-2-Iodo-1,3-bis[1-(mesitylcarbamoyl)ethoxy]benzene ≥98.0% (by HPLC)
  TCIAI0807-200MG
 :  TCI
 :  
(R,R)-2-Iodo-1,3-bis[1-(mesitylcarbamoyl)ethoxy]benzene ≥98.0% (by HPLC)
  TCIAI0807-200MG
 :  TCI
 :  

 

  • Pk:
    200 mg
  • Seq:
    2530

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

 

 

 :
 :614.52 g/mol
 :Refrigerator
 :1226896-38-3
 :C₃₀H₃₅IN₂O₄