3-Isoamyl-6-methyl-2-heptyl myristate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:88332-30-3
:
Formula:C₂₇H₅₄O₂
:410.73 g/mol
Flash Pt:205°C
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together