
(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:131457-46-0
:
Formula:C₂₁H₂₂N₂O₂
MDL Number:MFCD00192245
:334.42 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 95.0 %
- Specific rotation [a]20/D:-151 to -173 deg (C=1, EtOH)