2,3-O-Isopropylidene-D-erythronolactone ≥98.0% (by GC)
TCIAI0454-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:66.0 to 70.0 deg-C
- Specific rotation [a]20/D:-116 to -123 deg (C=1, H2O)
:
:158.15 g/mol
:25581-41-3
:C₇H₁₀O₄
:MFCD00134440