
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAH1652-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:64.0 to 68.0 deg-C
Specification Test Results
About this item
:
:184.19 g/mol
:GHS07
CAS Number:88335-93-7
Formula:C₉H₁₂O₄
MDL Number:MFCD00075490