9-Hexyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:871696-12-7
:
Formula:C₃₀H₄₃B₂NO₄
MDL Number:MFCD30187269
:503.30 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Melting point:221.0 to 225.0 deg-C