(2S,4R)-Methyl-4-hydroxypyrrolidine-2-carboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:40216-83-9
:
Formula:C₆H₁₁NO₃·HCl
MDL Number:MFCD00080855
:181.6 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:-27.0 to -31.0 deg (C=4,H2O)