
2-Hydroxy-3-methylpyrazine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:19838-07-4
:
Formula:C₅H₆N₂O
MDL Number:MFCD01646120
:110.11 g/mol
EINECS:243-362-7
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:148.0 to 152.0 deg-C