Order Entry
Norway
ContactUsLinkComponent
 
4,4,4,4',4',4'-Hexafluoro-DL-valine ≥98.0% (by titrimetric analysis)
  TCIAH1427-200MG
 :  TCI
 :  
undefined
4,4,4,4',4',4'-Hexafluoro-DL-valine ≥98.0% (by titrimetric analysis)
  TCIAH1427-200MG
 :  TCI
 :  

 

  • Pk:
    200 mg
  • Seq:
    2530

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :225.09 g/mol
 :240-207-5
 :16063-80-2
 :C₅H₅F₆NO₂
 :MFCD00014349