
trans-10-Hydroxy-2-decenoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14113-05-4
:
Formula:C₁₀H₁₈O₃
MDL Number:MFCD00204506
:186.25 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:64.0 to 68.0 deg-C