Order Entry
Norway
ContactUsLinkComponent
 
 
3-Hydroxy-1-adamantane carboxylic acid ≥97.0% (by GC, titration analysis)
 :  TCI
 
 
3-Hydroxy-1-adamantane carboxylic acid ≥97.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization titration):
    min. 97.0 %
  • Melting point:
    200.0 to 205.0 deg-C